Anti-Helicobacter pylori Mouse Monoclonal Antibody (Biotin) [clone: HP/212]
Leverantör: Biotium
BNCB0212-500 BNCB0212-100
BNCB0212-500EA 6500 SEK
BNCB0212-500 BNCB0212-100
Anti-Helicobacter pylori Mouse Monoclonal Antibody (Biotin) [clone: HP/212]
Antikroppar
The spiral shaped bacteriumHelicobacter pyloriis strongly associated with inflammation of the stomach and is also implicated in the development of gastric malignancy.H. pyloriis known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcinoma and low-grade lymphoma of mucosa associated lymphoid tissue in the stomach.This antibody stains the individual H. pylori bacterium when it presents on the surface of the epithelium or in the cytoplasm of the epithelial cells in biopsy tissue sections from the antrum and body of the stomach.
Type: Primary
Antigen: Helicobacter pylori
Clonality: Monoclonal
Clone: HP/212
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Helicobacter pylori