Anti-KRT17 Mouse Monoclonal Antibody (Purified) [clone: E3]

Leverantör: Biotium

BNUB0158-500 BNUB0158-100
BNUB0158-500EA 5625 SEK
BNUB0158-500 BNUB0158-100
Anti-KRT17 Mouse Monoclonal Antibody (Purified) [clone: E3]
Antikroppar
Cytokeratin 17 (CK17) is normally expressed in the basal cells of complex epithelia but not in stratified or simple epithelia. Antibody to CK17 is an excellent tool to distinguish myoepithelial cells from luminal epithelium of various glands such as mammary, sweat and salivary. CK17 is expressed in epithelial cells of various origins, such as bronchial epithelial cells and skin appendages. It may be considered as anepithelial stem cellmarker because CK17 Ab marks basal cell differentiation. CK17 is expressed in SCLC much higher than in LADC. Eighty-five percent of the triple negative breast carcinomas immunoreact with basal cytokeratins including anti-CK17. Also important is that cases of triple negative breast carcinoma with expression of CK17 show an aggressive clinical course. The histologic differentiation of ampullary cancer, intestinal vs. pancreatobiliary, is very important for treatment. Usually anti-CK17 and anti-MUC1 immunoreactivity represents pancreatobiliary subtype whereas anti-MUC2 and anti-CDX-2 positivity defines intestinal subtype.

Type: Primary
Antigen: KRT17
Clonality: Monoclonal
Clone: E3
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Cow, Goat, Human, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR