Anti-Liver Canuliculi Mouse Monoclonal Antibody (CF647) [clone: HSA98]

Leverantör: Biotium
BNC470098-500 BNC470098-100
BNC470098-500EA 6500 SEK
BNC470098-500 BNC470098-100
Anti-Liver Canuliculi Mouse Monoclonal Antibody (CF647) [clone: HSA98]
Antikroppar
Monoclonal antibodies (MAb's) to liver cell processes are useful in the identification of hepatic carcinomas and normal organ structures. MAb HSA98 binds to human hepatocytes and the majority of human hepatocellular carcinomas (HCC's). In frozen sections, it stains hepatic cells and may be used as a marker of the liver. Cell preparations of hepatocellular carcinoma biopsies or cell lines are found to bind HSA98 on the cell surface. This MAb stains liver hepatocytes in frozen human liver sections and is positive on the cell surface of human liver carcinomas.

Type: Primary
Antigen: Liver Canuliculi
Clonality: Monoclonal
Clone: HSA98
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR