Anti-Vtg Mouse Monoclonal Antibody [clone: ND-1E8]
Leverantör: Cayman Chemical
10007143-100 10007143-500
CAYM10007143-100EA 10775 SEK
CAYM10007143-100 CAYM10007143-500
Anti-Vtg Mouse Monoclonal Antibody [clone: ND-1E8]
Antikroppar
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: ND-1E8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: American Alligator