Anti-IgG lambda Mouse Monoclonal Antibody [clone: ICO-106]

Leverantör: US Biological
138563 138563-100
USBI138563EA 4550 SEK
USBI138563 USBI138563-100
Anti-IgG lambda Mouse Monoclonal Antibody [clone: ICO-106]
Antikroppar
Recommended dilution: Immunofluorescence: 1-2?g/ml; ;Western Blot: 0.5-1 ?g/ml; Immunoprecipitation: 1-2?g/500 ?g Immunochemistry: 0.5-1 ?g/ml

Type: Secondary
Antigen: IgG lambda
Clonality: Monoclonal
Clone: ICO-106
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR