Anti-APRIL Rabbit Polyclonal Antibody (Biotin)

Leverantör: US Biological
143550 143550-50
USBI143550EA 3775 SEK
USBI143550 USBI143550-50
Anti-APRIL Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 0.25-1 ?g/ml

Type: Primary
Antigen: APRIL
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR