Anti-Shh Rat Monoclonal Antibody

Leverantör: US Biological
S1012-96P-100 S1012-96P-500
USBIS1012-96P-100EA 9975 SEK
USBIS1012-96P-100 USBIS1012-96P-500
Anti-Shh Rat Monoclonal Antibody
Antikroppar
Recommended dilution: ELISA: 2-8?g/ml; ELISA: 0.5-2?g/ml; Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: Shh
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR