Anti-ENG Mouse monoclonal antibody unconjugated [clone: 3A9]
Leverantör: Novus Biologicals
NBP2-22122 NBP2-22122SS
NOVUNBP2-22122EA 7425 SEK
NOVUNBP2-22122 NOVUNBP2-22122SS
Anti-ENG Mouse monoclonal antibody unconjugated [clone: 3A9]
Antikroppar
Type: Primary
Antigen: ENG (endoglin)
Clonality: monoclonal
Clone: 3A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human