Anti-MLXIPL Rabbit Polyclonal Antibody

Leverantör: Novus Biologicals
NB400-136 NB400-136SS
NOVUNB400-136EA 6600 SEK
NOVUNB400-136 NOVUNB400-136SS
Anti-MLXIPL Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: MLXIPL (MLX interacting protein-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR