Anti-PLTP Rabbit Polyclonal Antibody

Leverantör: Novus Biologicals
NB400-106 NB400-106SS
NOVUNB400-106EA 6500 SEK
NOVUNB400-106 NOVUNB400-106SS
Anti-PLTP Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: PLTP (phospholipid transfer protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR