Anti-HIP1 Mouse Monoclonal Antibody [clone: 1B11]

Leverantör: Novus Biologicals
NB300-204 NB300-204SS
NOVUNB300-204EA 6600 SEK
NOVUNB300-204 NOVUNB300-204SS
Anti-HIP1 Mouse Monoclonal Antibody [clone: 1B11]
Antikroppar
Type: Primary
Antigen: HIP1 (huntingtin interacting protein 1)
Clonality: monoclonal
Clone: 1B11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR