Anti-NSP 5 alpha 3 alpha Rabbit Polyclonal Antibody

Leverantör: Novus Biologicals
NB100-517 NB100-517SS
NOVUNB100-517EA 6450 SEK
NOVUNB100-517 NOVUNB100-517SS
Anti-NSP 5 alpha 3 alpha Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: NSP 5 alpha 3 alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR