Anti-IgG Goat Polyclonal Antibody (CF™ 543)

Leverantör: Biotium
20311 20311-1
20311.EA 4175 SEK
20311. 20311-1
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Antikroppar
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR