Anti-IgG2a Goat Polyclonal Antibody (CF™ 555)

Leverantör: Biotium

20257 20257-1
20257.EA 4325 SEK
20257. 20257-1
Anti-IgG2a Goat Polyclonal Antibody (CF™ 555)
Antikroppar
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR