Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Leverantör: Biotium
20232 20232-1
20232.EA 4025 SEK
20232. 20232-1
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antikroppar
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit