Anti-IgG Donkey Polyclonal Antibody (CF™ 568)
Leverantör: Biotium
20098 20098-1
20098.EA 4175 SEK
20098. 20098-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 568)
Antikroppar
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit