Anti-IgG Donkey Polyclonal Antibody (CF™ 555)

Leverantör: Biotium

20038 20038-1
20038.EA 4025 SEK
20038. 20038-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 555)
Antikroppar
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR