Anti-IgG Rabbit Polyclonal Antibody (CF™ 555)
Leverantör: Biotium
20031 20031-1
20031.EA 3375 SEK
20031. 20031-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 555)
Antikroppar
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat