Anti-PBK/SPK Rabbit Monoclonal Antibody [clone: EP2520Y]

Leverantör: Abcam
AB75987-100 AB75987-1ML
ABCAAB75987-100EA 7650 SEK
ABCAAB75987-100 ABCAAB75987-1ML
Anti-PBK/SPK Rabbit Monoclonal Antibody [clone: EP2520Y]
Antikroppar
Rabbit monoclonal [EP2520Y] to PBK/SPK.

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PBK/SPK
Clonality: Monoclonal
Clone: EP2520Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR