Anti-BCL2L1 Rabbit polyclonal antibody unconjugated

Leverantör: Biorbyt
ORB99382-100 ORB99382-20
BIRBORB99382-100EA 0 SEK
BIRBORB99382-100 BIRBORB99382-20
Anti-BCL2L1 Rabbit polyclonal antibody unconjugated
Antikroppar
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR