Anti-MAZ Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB128342-100 ORB128342-200
BIRBORB128342-100EA 0 SEK
BIRBORB128342-100 BIRBORB128342-200
Anti-MAZ Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: MAZ (MYC-associated zinc finger protein (purine-binding transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR