Anti-LRAT Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB128295-100 ORB128295-200
BIRBORB128295-100EA 0 SEK
BIRBORB128295-100 BIRBORB128295-200
Anti-LRAT Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: LRAT (lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR