Anti-Potassium Channel Kv3.2b Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB128249-100 ORB128249-200
BIRBORB128249-100EA 0 SEK
BIRBORB128249-100 BIRBORB128249-200
Anti-Potassium Channel Kv3.2b Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: Potassium Channel Kv3.2b
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR