Anti-LAT Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB127412-100 ORB127412-200
BIRBORB127412-100EA 0 SEK
BIRBORB127412-100 BIRBORB127412-200
Anti-LAT Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR