Anti-IL11RA Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB125885-100 ORB125885-200
BIRBORB125885-100EA 0 SEK
BIRBORB125885-100 BIRBORB125885-200
Anti-IL11RA Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: IL11RA (interleukin 11 receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR