Anti-RMDN3 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB101821-100 ORB101821-200
BIRBORB101821-100EA 0 SEK
BIRBORB101821-100 BIRBORB101821-200
Anti-RMDN3 Rabbit Polyclonal Antibody
Antikroppar
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: RMDN3 (regulator of microtubule dynamics 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR