Anti-MAL and lymphocyte protein Rabbit Polyclonal Antibody
Leverantör: Biorbyt
ORB100788-100 ORB100788-200
BIRBORB100788-100EA 0 SEK
BIRBORB100788-100 BIRBORB100788-200
Anti-MAL and lymphocyte protein Rabbit Polyclonal Antibody
Antikroppar
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: MAL and lymphocyte protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now