Anti-MAL Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB100770-100 ORB100770-200
BIRBORB100770-100EA 0 SEK
BIRBORB100770-100 BIRBORB100770-200
Anti-MAL Rabbit Polyclonal Antibody
Antikroppar
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: MAL (mal, T-cell differentiation protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR