Anti-S100A2 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB100643-100 ORB100643-200
BIRBORB100643-100EA 0 SEK
BIRBORB100643-100 BIRBORB100643-200
Anti-S100A2 Rabbit Polyclonal Antibody
Antikroppar
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Primary
Antigen: S100A2 (S100 calcium binding protein A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR