Anti-ROM-K Rabbit Polyclonal Antibody
Leverantör: Biorbyt
ORB100146-100 ORB100146-200
BIRBORB100146-100EA 0 SEK
BIRBORB100146-100 BIRBORB100146-200
Anti-ROM-K Rabbit Polyclonal Antibody
Antikroppar
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200
Type: Primary
Antigen: ROM-K
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now