Anti-LCAT Rabbit Polyclonal Antibody
Leverantör: Biorbyt
ORB13538-100 ORB13538-200
BIRBORB13538-100EA 0 SEK
BIRBORB13538-100 BIRBORB13538-200
Anti-LCAT Rabbit Polyclonal Antibody
Antikroppar
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: LCAT (lecithin-cholesterol acyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat