Anti-ITGA2B Mouse Monoclonal Antibody (Biotin)

Leverantör: Biorbyt
ORB12056-100 ORB12056-500
BIRBORB12056-100EA 0 SEK
BIRBORB12056-100 BIRBORB12056-500
Anti-ITGA2B Mouse Monoclonal Antibody (Biotin)
Antikroppar
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR