Anti-Clara cell protein 16 Rabbit polyclonal antibody

Leverantör: Biorbyt
ORB10261-100 ORB10261-200
BIRBORB10261-100EA 0 SEK
BIRBORB10261-100 BIRBORB10261-200
Anti-Clara cell protein 16 Rabbit polyclonal antibody
Antikroppar
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Clara cell protein 16
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR