Anti-PLIN2 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB10045-100 ORB10045-200
BIRBORB10045-100EA 0 SEK
BIRBORB10045-100 BIRBORB10045-200
Anti-PLIN2 Rabbit Polyclonal Antibody
Antikroppar
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: PLIN2 (perilipin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR