Anti-RP1 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB2915-100 ORB2915-200
BIRBORB2915-100EA 0 SEK
BIRBORB2915-100 BIRBORB2915-200
Anti-RP1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: RP1 (retinitis pigmentosa 1 (autosomal dominant))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR