Anti-RTN4IP1 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
ORB2569-100 ORB2569-200
BIRBORB2569-100EA 0 SEK
BIRBORB2569-100 BIRBORB2569-200
Anti-RTN4IP1 Rabbit Polyclonal Antibody
Antikroppar
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: RTN4IP1 (reticulon 4 interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat