Anti-MAL Rabbit Polyclonal Antibody

Leverantör: Biorbyt
ORB2013-100 ORB2013-200
BIRBORB2013-100EA 0 SEK
BIRBORB2013-100 BIRBORB2013-200
Anti-MAL Rabbit Polyclonal Antibody
Antikroppar
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: MAL (mal, T-cell differentiation protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR