Genomic DNA isolation From tissues, E.Z.N.A.® MicroElute® Genomic DNA kit
Leverantör: OMEGA BIO-TEK
E.Z.N.A.® MicroElute® Genomic DNA kit provides a rapid and easy method for the isolation of genomic DNA from small sample volumes and sizes for consistent PCR and analytical test results. Using the reversible binding properties of a silica-based material, the speed of mini-column spin technology, and specifically formulated buffers, the system allows the purification of genomic DNA up to 60 kb. Samples are first lysed under denaturing conditions and then applied to the HiBind MicroElute® Column to bind DNA, while cellular debris, haemoglobin, and other proteins are effectively washed away. High quality DNA can be eluted with 10 to 15 µl sterile deionised water or elution buffer from the MicroElute® column.
- Purify genomic DNA from small sample volumes and small amount of cells
- High quality genomic DNA purification from variety of samples
- No phenol/chloroform extractions
- Purified DNA suitable for most downstream applications