Chemistry vacuum pump, NT series
Leverantör: VACUUBRAND
743301 20743300 20743500 743501 743302 743602 743601 20743600 743502
VACU743301EA 95575 SEK
VACU743301 VACU20743300 VACU20743500 VACU743501 VACU743302 VACU743602 VACU743601 VACU20743600 VACU743502
Chemistry vacuum pump, NT series
Pumpar Membranpumpar
Compact, powerful pump gives ideal solution for continuous, oil-free pumping of corrosive gases and vapors with exceptional high pumping speed. Eight-cylinder NT pump features quiet operation with smooth, easy-to-clean surfaces.
  • High chemical and vapour resistance
  • Very high pumping speed even at low vacuum levels
  • Long diaphragm life with maintenance free drive system
  • Ultimate vacuum even with gas ballast
  • PTFE sandwich diaphragms for increased reliability and extended operating life
  • All internal parts are made of chemically resistant fluoroplastics
  • Noise
More Product Information