Anti-Trk-Fused Gene Protein Rabbit Polyclonal Antibody

Leverantör: US Biological
T8662-80
USBIT8662-80EA 6575 SEK
USBIT8662-80
Anti-Trk-Fused Gene Protein Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Trk-Fused Gene Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR