Anti-RLBP1 Mouse Monoclonal Antibody [clone: 13A14]

Leverantör: US Biological
R1659-90
USBIR1659-90EA 5400 SEK
USBIR1659-90
Anti-RLBP1 Mouse Monoclonal Antibody [clone: 13A14]
Antikroppar
Recommended dilution: Western Blot: 1:10000; Immunoprecipitation: 2?l

Type: Primary
Antigen: RLBP1 (retinaldehyde binding protein 1)
Clonality: monoclonal
Clone: 13A14
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR