Anti-DRD2 Rabbit Polyclonal Antibody

Leverantör: US Biological
D9000-06H
USBID9000-06HEA 7375 SEK
USBID9000-06H
Anti-DRD2 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: DRD2 (dopamine receptor D2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

SPECIFIKATIONER

Antigensymbol DRD2
Antikroppsnamn Dopamine receptor D2
Environmentally Preferable
Antikroppstyp Primär
Klonalitet Polyclonal
Konjugation Unconjugated
Reaktivitet
Värd Rabbit
Isotyp IgG
Western blot Yes
ImmunKemi Yes
Form lyophilized powder
Rening purified
Storage buffer 0,1ml of PBS which contains 10 mg/ml BSA.
Förvaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR