Anti-DGKI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverantör: US Biological
D3441-87A-HRP
USBID3441-87A-HRPEA 10275 SEK
USBID3441-87A-HRP
Anti-DGKI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: DGKI (diacylglycerol kinase, iota)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR