Anti-CNR2 Goat Polyclonal Antibody

Leverantör: US Biological
150211
USBI150211EA 9400 SEK
USBI150211
Anti-CNR2 Goat Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:16000; Western Blot: 2-3?g/ml

Type: Primary
Antigen: CNR2 (cannabinoid receptor 2 (macrophage))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR