Anti-CNR1 Goat Polyclonal Antibody

Leverantör: US Biological
150210
USBI150210EA 9400 SEK
USBI150210
Anti-CNR1 Goat Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:32000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: CNR1 (cannabinoid receptor 1 (brain))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR