Anti-TLN1 Mouse Monoclonal Antibody [clone: 1A11]

Leverantör: US Biological
134483
USBI134483EA 7625 SEK
USBI134483
Anti-TLN1 Mouse Monoclonal Antibody [clone: 1A11]
Antikroppar
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: TLN1 (talin 1)
Clonality: monoclonal
Clone: 1A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR