Anti-BAD Rabbit Polyclonal Antibody

Leverantör: US Biological
032405
USBI032405EA 7750 SEK
USBI032405
Anti-BAD Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Dot blot 1:500

Type: Primary
Antigen: BAD (BCL2-associated agonist of cell death)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR