Anti-IgG (H+L) Rabbit Polyclonal Antibody

Leverantör: Rockland Immunochemicals
809-4102
ROCK809-4102EA 5275 SEK
ROCK809-4102
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Antikroppar
Suitable for highly specific immunological methods requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:30,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR