Anti-IgG (H+L) Goat Polyclonal Antibody (Cy2®)

Leverantör: Rockland Immunochemicals
710-111-032
ROCK710-111-032EA 5850 SEK
ROCK710-111-032
Anti-IgG (H+L) Goat Polyclonal Antibody (Cy2®)
Antikroppar
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR