Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)

Leverantör: Rockland Immunochemicals
612-4326
ROCK612-4326EA 4250 SEK
ROCK612-4326
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)
Antikroppar
Secondary Rabbit Anti-IgG (H&L) Reacts with Rat

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR